콘텐츠로 건너뛰기
Merck
  • Molecular Cloning and Effects of Tm14-3-3ζ-Silencing on Larval Survivability Against E. coli and C. albicans in Tenebrio molitor.
Molecular Cloning and Effects of Tm14-3-3ζ-Silencing on Larval Survivability Against E. coli and C. albicans in Tenebrio molitor.
Genes (2018-07-04)
Jeong Hwan Seong, Yong Hun Jo, Gi Won Seo, Soyi Park, Ki Beom Park, Jun Ho Cho, Hye Jin Ko, Chang Eun Kim, Bharat Bhusan Patnaik, Sung Ah Jun, Yong Seok Choi, Young Wook Kim, In Seok Bang, Yong Seok Lee, Yeon Soo Han
초록
The 14-3-3 family of proteins performs key regulatory functions in phosphorylation-dependent signaling pathways including cell survival and proliferation, apoptosis, regulation of chromatin structure and autophagy. In this study, the zeta isoform of 14-3-3 proteins (designated as Tm14-3-3ζ) was identified from the expressed sequence tags (ESTs) and RNA sequencing (RNA-Seq) database of the coleopteran pest, Tenebrio molitor. Tm14-3-3ζ messenger RNA (mRNA) is expressed at higher levels in the immune organs of the larval and adult stages of the insect and exhibit almost five-fold induction within 3 h post-infection of the larvae with Escherichia coli and Candida albicans. To investigate the biological function of Tm14-3-3ζ, a peptide-based Tm14-3-3ζ polyclonal antibody was generated in rabbit and the specificity was confirmed using Western blot analysis. Immunostaining and confocal microscopic analyses indicate that Tm14-3-3ζ is mainly expressed in the membranes of midgut epithelial cells, the nuclei of fat body and the cytosol of hemocytes. Gene silencing of Tm14-3-3ζ increases mortality of the larvae at 7 days post-infection with E. coli and C. albicans. Our findings demonstrate that 14-3-3ζ in T. molitor is essential in the host defense mechanisms against bacteria and fungi.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline