콘텐츠로 건너뛰기
Merck
  • Gas chromatography-mass spectrometry determination of polycyclic aromatic hydrocarbons in baby food using QuEChERS combined with low-density solvent dispersive liquid-liquid microextraction.
Gas chromatography-mass spectrometry determination of polycyclic aromatic hydrocarbons in baby food using QuEChERS combined with low-density solvent dispersive liquid-liquid microextraction.
Food chemistry (2018-04-07)
Mateus Henrique Petrarca, Helena Teixeira Godoy
초록
A sensitive GC-MS method is reported for the determination of twelve polycyclic aromatic hydrocarbons (PAHs) in baby food. The sample preparation involves QuEChERS extraction combined with low-density solvent dispersive liquid-liquid microextraction (LDS-DLLME) and ultra-low temperature (-80 °C). Plackett-Burman screening design was employed to identify the main sample preparation variables that affect the extraction efficiency, such as the volume of toluene used in LDS-DLLME. The suitability of proposed method was verified by analytical selectivity, linearity in solvent and matrix-matched calibration curves and adequate recoveries (72-112%) and precision (RSD values ≤11%), under repeatability and within-laboratory reproducibility conditions. High analytical sensitivity was achieved for the monitoring of PAHs at the strict limit of 1 µg kg-1 fixed by the European Commission for baby foods. The validated method was applied to thirty-two commercial baby food samples, and the investigated PAHs were not detected in any sample.
MATERIALS
제품 번호
브랜드
제품 설명
Dibenzo[a,e]pyrene, BCR®, certified reference material
Sigma-Aldrich
Benzo[b]fluoranthene, 98%