- Induction of human cytochrome P450 3A4 by the irreversible myeloperoxidase inactivator PF-06282999 is mediated by the pregnane X receptor.
Induction of human cytochrome P450 3A4 by the irreversible myeloperoxidase inactivator PF-06282999 is mediated by the pregnane X receptor.
Xenobiotica; the fate of foreign compounds in biological systems (2017-07-08)
Jamie E Moscovitz, Zhiwu Lin, Nathaniel Johnson, Meihua Tu, Theunis C Goosen, Yan Weng, Amit S Kalgutkar
PMID28685622
초록
1. 2-(6-(5-Chloro-2-methoxyphenyl)-4-oxo-2-thioxo-3,4-dihydropyrimidin-1(2H)-yl) acetamide (PF-06282999) is a member of the thiouracil class of irreversible inactivators of human myeloperoxidase enzyme and a candidate for the treatment of cardiovascular disease. PF-06282999 is an inducer of CYP3A4 mRNA and midazolam-1'-hydroxylase activity in human hepatocytes, which is consistent with PF-06282999-dose dependent decreases in mean maximal plasma concentrations (C
MATERIALS