Merck
Center Drain (CD) screw thread vials, heavy wall, with caps and PTFE/silicone septa volume 5 mL, clear glass vial, with graduations, O.D. × H 20 mm × 61 mm, pkg of 12 ea
Center Drain (CD) screw thread vials, heavy wall, with caps and PTFE/silicone septa
Center Drain (CD) screw thread vials, heavy wall, with caps and PTFE/silicone septa volume 5 mL, clear glass vial, with graduations, O.D. × H 20 mm × 61 mm, pkg of 12 ea
Products
제품 카테고리
브랜드
OD(mm)
용적(ml)
뚜껑 유형
색상
바이알 병 재질
바이알 병 색상
포장 단위