콘텐츠로 건너뛰기
Merck
1-(4-(2,4-DIPENTYL-PHENOXY)-BUTYL)-3-METHYL-PYRIDINIUM, BROMIDE AldrichCPR

1-(4-(2,4-DIPENTYL-PHENOXY)-BUTYL)-3-METHYL-PYRIDINIUM, BROMIDE

1-(4-(2,4-DIPENTYL-PHENOXY)-BUTYL)-3-METHYL-PYRIDINIUM, BROMIDE AldrichCPR
Linear Formula:
C26H40BrNO
CAS Number:
Molecular Weight:
462.519

Products

화학식량
특수 등급
Showing 1-3 of 3