제품 이름
GenElute™-E Single Spin DNA Cleanup Kit Reagents and consumables for clean-up of pre-purified DNA. 10, 50 or 250 purifications.
제품 번호
ec600
제품 브랜드
sigma
CAS 번호
Molecular Weight
테스트 | 규격 |
|---|---|
| Void OD A230 | 0.1 |
| Eluat OD A230 | 0.07 |
| Resin concentration(mL) | 6.75 mL |
| Salt depletion/conductivity [% or µS/cm] | 96% / <200 µS/cm |
| PCR(FAM) | Ct 29 |
| PCR(VIC) | Ct 29 |
| DNA recovery rate(%) | 0.763 |
| Agarose gel | Depletion visible maximum in 300bp band |
| pH | pH 8.5 |