모든 사진(1)
About This Item
실험식(Hill 표기법):
C9H10INO2
CAS Number:
Molecular Weight:
291.09
MDL number:
UNSPSC 코드:
12352200
SMILES string
NC(Cc1ccc(I)cc1)C(O)=O
가장 최신 버전 중 하나를 선택하세요:
Dirk Hellwig et al.
European journal of nuclear medicine and molecular imaging, 35(1), 24-31 (2007-09-12)
Radioactive amino-acids accumulate in gliomas even with an intact blood-brain-barrier. L-3-[(123)I]-iodo-alpha-methyl-tyrosine (IMT) is well established for SPECT imaging of gliomas. Recently, we introduced p-[(123)I]-iodo-L-phenylalanine (IPA) for the characterisation of brain lesions. This study compares both tracers in glioma patients. Eleven
[Metabolic studies of p-iodophenylalanine in the pancreas: a gateway to the development of radioiodinated amino acids for functional diagnosis].
K Kawai et al.
Kaku igaku. The Japanese journal of nuclear medicine, 25(11), 1263-1270 (1988-11-01)
Veerle Kersemans et al.
European journal of nuclear medicine and molecular imaging, 33(8), 919-927 (2006-03-31)
In vitro in the R1M cell model and in vivo in the R1M tumour-bearing athymic model, both [(123)I]-2-iodo-L: -phenylalanine and [(123)I]-2-iodo-D: -phenylalanine have shown promising results as tumour diagnostic agents for SPECT. In order to compare these two amino acid
R Karthikraj et al.
Rapid communications in mass spectrometry : RCM, 26(11), 1385-1391 (2012-05-05)
In chiral differentiation by mass spectrometry, use of a single reference that differentiates various classes of compounds including drugs is ideal, but so far there are no such reports in the literature. We have successfully used iodo-substituted amino acids for
B Chevrier et al.
European journal of biochemistry, 237(2), 393-398 (1996-04-15)
The structure of the complex of Aeromonas proteolytica aminopeptidase, a two-zinc exopeptidase, with the inhibitor p-iodo-D-phenylalanine hydroxamate has been determined by X-ray crystallography. Refinement of the structure, which includes 220 water molecules, using data at 0.80-0.23-nm resolution resulted in a
자사의 과학자팀은 생명 과학, 재료 과학, 화학 합성, 크로마토그래피, 분석 및 기타 많은 영역을 포함한 모든 과학 분야에 경험이 있습니다..
고객지원팀으로 연락바랍니다.