콘텐츠로 건너뛰기
Merck
모든 사진(1)

문서

ERD-083

Supelco

Diisopropyl methylphosphonate solution

1000 μg/mL in methanol, ampule of 1.2 mL, certified reference material, Cerilliant®

로그인조직 및 계약 가격 보기


About This Item

실험식(Hill 표기법):
C7H17O3P
CAS Number:
Molecular Weight:
180.18
EC Number:
MDL number:
UNSPSC 코드:
41116107
PubChem Substance ID:

Grade

certified reference material

특징

Snap-N-Spike®/Snap-N-Shoot®

포장

ampule of 1.2 mL

제조업체/상표

Cerilliant®

농도

1000 μg/mL in methanol

형식

single component solution

저장 온도

−20°C

SMILES string

O=P(OC(C)C)(C)OC(C)C

InChI

1S/C7H17O3P/c1-6(2)9-11(5,8)10-7(3)4/h6-7H,1-5H3

InChI key

WOAFDHWYKSOANX-UHFFFAOYSA-N

법적 정보

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

픽토그램

FlameSkull and crossbonesHealth hazard

신호어

Danger

유해 및 위험 성명서

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup


시험 성적서(COA)

제품의 로트/배치 번호를 입력하여 시험 성적서(COA)을 검색하십시오. 로트 및 배치 번호는 제품 라벨에 있는 ‘로트’ 또는 ‘배치’라는 용어 뒤에서 찾을 수 있습니다.

이 제품을 이미 가지고 계십니까?

문서 라이브러리에서 최근에 구매한 제품에 대한 문서를 찾아보세요.

문서 라이브러리 방문

Edward J Calabrese
Regulatory toxicology and pharmacology : RTP, 38(3), 260-268 (2003-11-19)
This analysis retrospectively assessed the likelihood that medical intervention masked effects of diisopropylmethylphosphonate (DIMP) on female mink. Bucci et al. [Two-generation reproductive study in mink fed DIMP. Final report. Study no. TP-001. Prepared by Pathology Associates International, Jefferson, AR, USA
T J Bucci et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 22(2), 220-230 (1994-02-01)
Diisopropyl methylphosphonate (DIMP), produced during manufacture of the chemical agent GB (Sarin), is a groundwater contaminant at Rocky Mountain Arsenal, Colorado. DIMP was fed for 90 days to dark brown "Ranch Wild" mink housed under controlled indoor conditions. One-year-old mink
Thomas J Bucci et al.
Reproductive toxicology (Elmsford, N.Y.), 17(3), 327-334 (2003-05-22)
Two generations of "Ranch Wild" mink (Mustela vison) were fed the organophosphate diisopropyl methylphosphonate (DIMP) at 0, 150, 450, or 1250ppm, to determine potential toxicity to the dams. Chemical, hematologic, necropsy, and microscopic examinations were performed on all parental animals
D M McPhillips et al.
Journal of biomedical materials research, 17(6), 993-1002 (1983-11-01)
A series of porous polymeric sorbents and activated carbon were used to remove diisopropyl methylphosphonate (DIMP) from human plasma and normal saline. The sorptive capacities of the commercially available sorbents Amberlite XAD-4, XAD-2, XN1010, and XE348, and Calgon 400 were
H L Snodgrass et al.
Journal of toxicology and environmental health, 36(4), 367-376 (1992-08-01)
Diisopropyl methylphosphonate (DIMP) has been identified as a groundwater contaminant on or near sites of former chemical warfare production facilities. The material is a by-product of GB (or Sarin) manufacture and does not occur naturally in the environment. The present

자사의 과학자팀은 생명 과학, 재료 과학, 화학 합성, 크로마토그래피, 분석 및 기타 많은 영역을 포함한 모든 과학 분야에 경험이 있습니다..

고객지원팀으로 연락바랍니다.