모든 사진(1)
Key Documents
WHA10447900
Whatman® Minifold® I 96 well dot-blot array system
Minifold I dot-blot system, complete system, 96 well (Acrylic) includes manifold apparatus, 5 x Protran BA85 sheets, 5 x 3 mm Chr sheets, 1/pk
동의어(들):
96 well dot blot systems, Z613932, complete dot blot system, dot blot systems, dot blotting systems
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41105300
NACRES:
NB.22
포장
pack of 1 (complete system)
제조업체/상표
Whatman 10447900
Whatman Article No. 28415657 (US reference)
일반 설명
Whatman® Minifold I Dot-Blot System
Minifold® I Dot-Blot System - 96 Well
  • Superior 96 Well Manifolds for Proteins and Nucleic Acids: Dot-Blot Array Systems
  • The Minifold I system consists of four basic components: sample well plate, filter support plate, vacuum plenum, and metal clamping plate. The sample well plate is available in three configurations for producing spots, dots or slots.
  • Novel O-ring design – ensures discrete dot formation without leakage of samples by cross lateral flow.
  • Generates even, uniform dots that eliminate uneven test areas resulting from manual sample application.
기타 정보
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
법적 정보
Minifold is a registered trademark of Whatman International Ltd.
Whatman is a registered trademark of Cytiva
시험 성적서(COA)
제품의 로트/배치 번호를 입력하여 시험 성적서(COA)을 검색하십시오. 로트 및 배치 번호는 제품 라벨에 있는 ‘로트’ 또는 ‘배치’라는 용어 뒤에서 찾을 수 있습니다.