Merck
HomeProtein PurificationAmicon® Pro Purification System – Protein Purification
Amicon® Pro Purification System – Protein Purification
The Amicon® Pro Purification System is an all-in-one centrifugal device for the protein purification workflow. Traditional protein purification is a long process with many steps and multiple tubes, often resulting in protein degradation and loss. Avoid the risks associated with sample transfer and reduce hands-on time when you bind, wash, elute and/or concentrate your protein in the all-in-one Amicon® Pro purification system. Regardless of your workflow, this virtually touchless device delivers consistent, accurate sample preparation, resulting in reliable recovery, uncompromised purity and easy data generation.
The Amicon® Pro Purification System Workflow
Bind, wash, elute and/or concentrate your protein in this all-in-one centrifugal device.
The Amicon® Pro Purification System Workflow
Elute, Don't Dilute
Excellent Recovery, Regardless of Scale
elute dont elute
Figure 1.While three elution steps were required for the smaller spin columns (left), a single spin in the Amicon® Pro device was sufficient to recover > 90% of resin-bound protein in an average of 45 µL final volume*.
Excellent recovery
Figure 2.Protein recovered increases linearly with respect to packed resin volume, offering flexibility in experimental scale. Whether you are using a few microliters or a milliliter of resin, Amicon® Pro purification yields consistent recovery.
Compare the Amicon® Pro Workflow with Traditional Methods
Purification with buffer exchange/concentrationPurification only
0.5 mL spin column + concentration deviceAmicon® Pro Purification System0.5 mL gravity columnAmicon® Pro Purification System
Process time67 minutes29 minutesa125 minutesb65 minutesa
Steps required126186
Sample transfers103-8 (# of fractions collected)0
a Process time excludes 60 minute incubation for both protocols.
b Gravity column process includes column setup, purification and assessment of eluted fractions.
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch