Merck
HomeBiosensors & BioimagingSelf healable PEDOT based film hydrogel and OECT devices
Self-healable PEDOT-based film, hydrogel, and OECT devices for bioelectronics
Meijing Wang, Jinsil Kim, Floriane Miquet-Westphal, Xin Zhou, Pierre Kateb, Jiaxin Fan, Fabio Cicoira
Department of Chemical Engineering, Polytechnique Montréal, H3T 1J4, Montréal (QC), CANADA
Material Matters™ 2024, 19.1
Introduction
In recent years, bioelectronics has made remarkable progress in developing wearable and implantable devices that can monitor and interact with biological systems due to the development of conducting polymer-based self-healing materials.1 The human body is composed of living organisms characterized by the presence of bioelectricity, which is produced by the flow of ions across biological membranes. Measuring these electrical signals allows for the evaluation of different physiological parameters of the body, such as the electroencephalogram (EEG) for the brain, the electrocardiogram (ECG) for the heart, and the electromyography (EMG) for muscle responses.2 With its customized and non-invasive healthcare options, bioelectronics promises to transform medical practices, providing new ways to diagnose and treat various illnesses.3
Materials used in bioelectronics are required to mimic the properties of biological tissues. In this context, the current devices made of rigid inorganic materials may experience damage or degradation from physical stress and chemical interactions. On the other hand, soft organic polymers combine mechanical flexibility and high electrical conductivity, allowing for the development of more advanced bioelectronics.4 Another challenge in bioelectronics is the requirement of conductive materials to maintain their performance for extended periods. Self-healing conductors have gained attention due to their ability to repair damages in their conducting structure, thus enhancing the durability and longevity of electronic devices by restoring electrical conductivity. This makes them a potential solution to address the issue at hand. Self-healing can occur through various mechanisms, including chemical reactions, physical interactions (mechanical or thermal), or shape-memory effects.5 In bioelectronics, self-healing materials are beneficial as they can repair themselves in dynamic biological environments.6
The organic conducting polymer, poly(3,4-ethylenedioxythiophene) (PEDOT) doped with polystyrene sulfonate (PSS), has been widely used for developing self-healable conductors due to its ease of processing in aqueous media, high electrical conductivity, and self- healing properties.1 Herein, we discuss the recent advances in self- healable conductors based on PEDOT:PSS (Cat. Nos. 483095, 900208, 900181) and organic electrochemical transistor (OECT) devices based on these materials.
Self-healable PEDOT:PSS-based Films
The conjugated structure of organic conducting polymers, composed of carbon atoms with alternating single and double bonds along the polymer backbone, facilitates electron delocalization along the entire chain and leads to electrical conductivity. The conductivity of the conjugated system can be enhanced by doping, such as PSS- doped PEDOT.7 In the PEDOT:PSS system, the conductivity can be further improved by incorporating conductivity enhancers, such as glycerol (Cat. No. G7893), sorbitol (Cat. No. 56755-M), ethylene glycol (EG, Cat. No. 102466), and dimethyl sulfoxide (DMSO, Cat. No. 276855).8 Pure conducting polymers generally exhibit poor mechanical properties, characterized by brittleness and fragility, making them susceptible to mechanical damage such as cracking or breaking. Therefore, they must be combined with other materials to improve their mechanical properties. Self-healing conductive materials have been obtained by blending conducting polymers with other polymers, such as polyvinyl alcohol (PVA, Cat. No. 341584), polyethylene glycol (PEG, Cat. No. 202398), TritonX-100 (Cat. No. X100), agarose, and others. In the next section, we will introduce the self-healing performance and mechanical properties of PEDOT:PS-based materials.
Our group has reported that PEDOT:PSS film can undergo water- induced healing.9 In this work, the PEDOT:PSS films were drop-cast onto a glass slide, and the current flow was monitored throughout the process. The current flow was interrupted when a cut was made intentionally with a razor blade. The current was recovered after a water droplet was placed on the damaged area (Figure 1A). The cutting and healing process was repeated in various regions of the film, and excellent healing efficiency of 100% and rapid response time of 150 ms were observed. The scanning electron microscopy (SEM) image of the damaged PEDOT:PSS film reveals a noticeable cut gap before water contact and mechanical gap recovery after healing (Figure 1B). Adding water to the separated PEDOT:PSS films may cause them to swell and increase in volume, enabling the filling of wounded areas and the restoration of conductivity. Based on the observation of the water-enabled rapid healing property of PEDOT:PSS thin films, wireless water sensors using water-healable PEDOT:PSS films were demonstrated.
In a successive work, we investigated the water-induced healing property of PEDOT:PSS-based films by studying the impact of 3-(glycidyloxypropyl)trimethoxysilane (GOPS, Cat. No. 440167) incorporation and the effect of sulfuric acid post-treatment.10 The healing performance can be tailored by adjusting the concentration of GOPS and the duration of sulfuric acid treatment. Increasing the volume ratio of GOPS to PEDOT:PSS from 0.5 to 1 resulted in a conductivity decrease from 350 S cm-1 to 300 S cm-1 and a healing efficiency reduction from 66% to 15%. While sulfuric acid post- treatment significantly improved the conductivity of PEDOT:PSS films, the healing efficiency decreased to 93%, 75%, and 70%, depending on the soaking time in strong acid (Figure 1E). It is reported that increasing GOPS content and sulfuric acid treatment duration influences the swelling of PSS, which was observed as a decrease in weight gain of the PEDOT:PSS films, resulting in deterioration of self-healing ability. In addition, we also explored PEDOT films with different dopants. We observed that inserting organic dopants, tosylate (Tos) and triflate (OTf), through chemical polymerization can lead to water-enabled healing properties. In contrast, the inorganic perchlorate ion (ClO4-) doped film obtained from electro-polymerization showed no healing ability.
When blended with a small amount (<10%) of certain additives, the PEDOT:PSS-based films exhibit autonomous healing without needing assistance from water. This exceptional capability is achieved by adjusting the film’s mechanical properties with these additives.11 This proposed strategy enhances the autonomous self- healing property typically observed only in wet PEDOT:PSS film. PEDOT:PSS mixed with low molecular weight PEG (200 or 400) have achieved autonomous healing and showed 100% healing efficiency with a 1 second response time (Figure 1C). With an increase in PEG molecular weight to 1500, the healing efficiency decreased to 80% (Figure 1C). We reported that the high molecular weight of PEG caused lower chain mobility, resulting in decreased viscoelastic behavior in the film. Adding PEG 400 to PEDOT:PSS decreased storage and loss modulus while increasing the elongation at break from 1% to 9%. Therefore, the composite exhibited viscoelastic behavior, and autonomous healing was achieved at a PEG concentration of 4 vol%.
Adding Triton X-100 to PEDOT:PSS can improve its viscoelastic properties, allowing for repetitive electrical current healing without external stimulation (Figure 1D). This film has been reported to have a stable electrical response during repetitive folding and stretching.12 The polymer blend of PEDOT:PSS and a tandem repeat protein derived from squid ring teeth (SRT) has exhibited electrical and mechanical healing abilities at 70 °C in wet conditions. The recovery was observed when 40 wt% SRT was added to PEDOT:PSS due to the interaction between PSS and SRT proteins at 70°C in the presence of water.13
This graph illustrates the water-induced electrical healing of PEDOT:PSS film. It presents a current versus time plot, demonstrating sharp drops in current when the film is cut, followed by a rapid return to near the original current level upon the addition of water. The graph visually captures the self-healing behavior of the film, showcasing its ability to recover from damage caused by cuts. This phenomenon is crucial for applications requiring robust and resilient materials, particularly in the field of flexible electronics and biomedical devices.
This set of scanning electron microscope (SEM) images reveals the damaged and healed areas of PEDOT:PSS film. The images depict the film at two different magnifications (20 um and 10 um), showcasing the structural changes before and after healing. Additionally, a diagram is included to visually represent the progression from a broken film to the film with water and, finally, the healed film. These images provide valuable insights into the microstructural changes that occur during the healing process, shedding light on the material's self-repair mechanisms and its potential for various practical applications.
This collection of three graphs presents the electrical healing capabilities of PEDOT:PSS films with different additives. Each graph showcases the current behavior of the films under specific conditions, demonstrating their response to cuts and subsequent healing. The first graph exhibits distinct lines related to 4% PEG-400 and 4% PEG-400/5% Glycerol, both showing sharp drops in current upon cutting and rapid healing to the original level. The second graph features a single line related to 4% PEG-200, while the third graph displays a line related to 4% PEG1500, showing slower healing to 80% of the original level. These graphs provide a comprehensive overview of the electrical healing properties of PEDOT:PSS films with various additives.
This graph depicts the repetitive self-healing behavior of PEDOT:PSS film, showcasing multiple cuts to the film and its ability to repeatedly return to the original current level. The graph presents a current versus time plot, highlighting sharp drops in current upon each cut, followed by the film's rapid self-repair to the original level. This visual representation effectively demonstrates the film's remarkable self-healing capabilities, indicating its potential for use in applications requiring sustained electrical conductivity and resilience to damage.
This set of graphs presents the healing efficiency and response time of PEDOT:PSS films in relation to various amounts of GOPS and sulfuric acid immersion time. The graphs reveal the impact of increased GOPS concentration and prolonged immersion time on the healing efficiency and response time of the films. Specifically, they demonstrate that higher GOPS concentration and longer immersion time lead to lower healing efficiency. These findings provide critical insights into the factors influencing the self-healing properties of PEDOT:PSS films and their behavior under different environmental conditions, offering valuable information for the development of advanced self-repairing materials.
Figure 1. A) Current vs. time measurement demonstrating water-induced electrical healing of PEDOT:PSS film. B) SEM images of the damaged and healed areas of PEDOT:PSS film. Reprinted with permission from reference 9, copyright 2017 Wiley Online Library. C) Current vs. time measurement with several cuts showing the different electrical healing capabilities of PEDOT:PSS films with 4% PEG-400, PEG-200, and PEG-1500. Reprinted with permission from reference 11, copyright 2020 Wiley Online Library. D) Current vs. time test on PEDOT:PSS/Triton X-100 system revealing the healing property. Reprinted with permission from reference 12, copyright 2016 Wiley Online Library. E) Healing efficiency and response time versus different amounts of GOPS and sulfuric acid immersion time of PEDOT:PSS films. Reprinted with permission from reference 10, copyright 2020 Wiley Online Library.
Self-healable Hydrogels
Conductive and self-healable hydrogels show great potential in bioelectronics and wearable electronics owing to their tunable mechanical and electrical properties, high water content, biocompatibility, and self-healing ability. Generally, self-healable hydrogels can be achieved either by utilizing pure conductive polymers (e.g., PEDOT:PSS) or, in most cases, by incorporating conductive materials (e.g., PEDOT:PSS, silver nanowires) into a matrix of self-healable polymers (e.g., PVA, poly(acrylamide) (PAAM), and gelatin).1
Our group reported self-healable PEDOT: PSS-based hydrogels obtained from mixing PVA, borax, and PEDOT:PSS screen printing ink (CleviosTM SV3, containing PEDOT:PSS, propylene glycol (PG, Cat. No. P4347), and diethylene glycol (DEG, Cat. No. H26456) (Figure 2A).14In this work, the as-prepared SV3/PVA hydrogels exhibited high adhesion (~2 N/cm2 on porcine skin), high plastic stretchability (>10000% strain), and a low compressive Young’s modulus (~4 kPa). In addition, the hydrogels present repeated self-healing abilities with a healing efficiency of ~100% due to the introduction of the ink into the PVA-borax system (Figure 2B). Epidermal electrodes fabricated using the PEDOT: PSS-based self- healing hydrogel exhibited excellent signal quality for ECG and EMG recording, which is comparable to that of commercial Ag/AgCl gel electrodes (Cat. Nos. BASMF2056, BASMF2052).14
The conductive hydrogels have been reported to be prone to environmental factors such as high or low temperatures, which impedes their practical application for long-term use. This is primarily due to water loss, which can adversely affect their mechanical and electrical properties. Water-retaining and anti- freezing organohydrogels have been developed to address this issue by incorporating anti-freezing agents, such as EG and glycerol, into the formulations.15–17 Self-healable organohydrogels obtained from PEDOT:PSS, PVA, and EG showed anti-freezing ability and high stretchability (200–900% strain). In addition, the gels exhibited self-healing behavior with a healing efficiency of ~85% (Figure 2C), which was attributed to the dynamic dissociation and re-association of crystalline domains and hydrogen bonds.18
This illustration visually depicts the step-by-step preparation of SV3/PVA hydrogel. In Step 1, a PVA solution and PEDOT:PSS paste are added to a vial and mixed to create a homogeneous mixture. Step 2 involves adding a borax solution to the vial, which leads to the formation of the SV3/PVA hydrogel. The illustration provides a clear and informative representation of the hydrogel preparation process, highlighting the essential steps involved in creating the homogeneous hydrogel material.
The graph on the left labeled b, presents a current versus time plot, demonstrating the electrical self-healing behavior of SV3/PVA hydrogel. It showcases multiple cuts to the hydrogel, each resulting in sharp drops in current, followed by repetitive self-healing to the original level. The graph effectively captures the remarkable self-repair capabilities of the hydrogel, indicating its potential for applications requiring sustained electrical conductivity and resilience to damage. The graph on the right labeled c, compares the tensile stress versus strain behavior of the original and healed organohydrogels. It visually demonstrates that the original gel can handle higher stress than the healed gel, indicating the impact of the healing process on the mechanical properties of the material. The graph provides valuable insights into the mechanical behavior of the organohydrogels before and after healing, shedding light on their potential for use in applications requiring robust and resilient materials with tailored mechanical properties.
Figure 2.A) Illustration of SV3/PVA hydrogel preparation. B) Current vs. time plots upon several cutting-healing cycles for the SV3/PVA hydrogel showing repeated electrical self-healing. Reprinted with permission from reference 14, copyright 2022 Elsevier. C) Tensile stress vs. strain plots of the original and healed organohydrogels. Reprinted with permission from reference 16, copyright 2017 Wiley Online Library.
Self-healable OECTs
OECTs, or three-terminal devices, have attracted increasing interest in bioelectronics. An OECT is comprised of source and drain electrodes connected by an ion-permeable conducting polymer channel (e.g., PEDOT:PSS) and a liquid or gel electrolyte in direct contact with the channel and a gate electrode. Although the electrodes are typically prepared by microfabrication, etching, and metal deposition,19 printing is a valuable alternative, especially for flexible and stretchable substrates.20,21 The conducting polymer channel can be deposited via various techniques, including spin coating, electrospinning, vapor phase polymerization, electropolymerization, and printing.22 Concerning the gating media, although aqueous electrolytes are commonly used, gel electrolytes, such as iongels and hydrogels prepared by spin coating or printing, have been explored owing to their promising potential in flexible and stretchable electronics.19,20 The operation of OECT utilizes the interaction between ions from the electrolyte and the channel material. This is achieved by controlling the gate voltage, which drives ions in/out of the channel and alters the doping state of the conducting polymer, leading to a change in the channel conductivity. Gating through an electrolyte allows the OECT to operate under 1 V with large amplification of signals owing to their high volumetric capacitance. The combination of these merits makes OECTs a suitable candidate for biological applications.22
OECTs have been explored to record electrophysiological signals in vivo for healthcare monitoring. For example, ECG signals can be easily acquired by putting an OECT on the skin. Furthermore, the amplified signals of electrooculography (EOG), EEG, and EMG can be recorded to monitor human eye movement, brain activity, and muscle movement.23 OECTs can also be directly interfaced invasively with organs to access their local signals. For example, PEDOT:PSS OECTs have been implanted in rat brains to record epileptic seizures, demonstrating the capability of detecting the signals from a single neuron and inducing localized stimulation by current injection in vivo. Further mapping of the brain was realized by employing soft OECT arrays.24 Another vital application, OECT transducers, performing as ultra-sensitive biosensors, have been broadly investigated to detect ions and metabolites, such as Na+, K+, glucose, and lactate. High sensitivity has been demonstrated in detecting dopamine, adrenaline, DNA (deoxyribonucleic acid), and bacteria.25
Self-healing materials have been integrated into various functional devices, including physical or chemical sensors, organic field- effect transistors, and energy storage devices. However, research on self-healing OECTs and their applications is still limited due to the lack of self-healing channel materials. Developing self-healable materials that are capable of maintaining their charge transport properties simultaneously remains a challenge.26
We reported a ground-breaking study on the first self-healing PEDOT:PSS OECTs. The devices exhibited remarkable healing properties, as evidenced by the preservation of transfer characteristics upon in-situ razor blade damage. This significant finding further highlights the potential of OECTs for long-term bioelectronic applications.9 Subsequent studies explored all- solid-state self-healing OECTs, incorporating surfactant-modified PEDOT:PSS channels and PVA hydrogel electrolytes. The device showed excellent performance with high transconductance, fast response, long-term stability, and ion-sensing behaviors (Figure 3A), making it an interesting candidate for developing practical bioelectronic applications.27 Additionally, PEDOT:PSS hydrogel fibers through syringe injection were investigated as a potential channel material for OECTs and demonstrated self-healing properties, suggesting promising potential for healable bioelectronics.28 Recent work has successfully combined the outstanding stretchability and self-healing properties of PEDOT:PSS films by adding a soft polymer, leading to the successful realization of an OECT array (Figure 3B). This development marks a significant advancement in the field of bioelectronic devices.29 Alternatively, incorporating hydrogel electrolytes has also been shown to endow OECTs with self-healing properties and has been demonstrated to mimic various synaptic functions.30
This schematic diagram illustrates a PEDOT:PSS-based organic electrochemical transistor (OECT) with a focus on its self-healing performance. It highlights the electrodes, polymer process channel, and hydrogel within the device. An additional diagram showcases the self-healing process, depicting the damaged PEDOT:PSS/TX polymer channel, the application of the hydrogel, and the subsequent healing. Additionally, a transfer curve is included to visually demonstrate the self-healing interaction within the device. These visuals provide an insightful representation of the self-healing capabilities of the OECT, offering a clear understanding of the device's potential for robust and resilient performance in various applications.
This photo presents arrays of PEDOT:PSS-based organic electrochemical transistors (OECTs), accompanied by an enlarged diagram highlighting the electrodes, channel, SEBS substrate, cation, anion, source, gate, and drain within the device. Additionally, an SEM image reveals the comparison between the damaged channel and the healed channel. Furthermore, a graph is included to display the gap recovery and restoration of transistor performance. These visuals provide comprehensive insights into the structure and self-healing properties of the OECTs, offering valuable information on the device's potential for applications requiring stretchable and self-healable electronic components.
Figure 3. A) Schematic diagram of PEDOT:PSS-based OECT with self-healable performance shown in transfer curve and self-healing process of channel film with the hydrogel on top. Reprinted with permission from reference 22, copyright 2020 American Chemical Society. B) Stretchable and self-healable PEDOT:PSS-based OECTs arrays showing gap recovery and transistor performance restoration. Reprinted with permission from reference 24, copyright 2022 Wiley Online Library.
Conclusion and Outlook
PEDOT:PSS has been demonstrated to be a self-healable conducting polymer. Self-healing PEDOT composite-based films, hydrogels, and devices, mainly OECTs, have been widely explored for various bioelectronic applications. Although significant progress has been achieved in materials development and application discovery, there are still challenges and new chances for self-healable bioelectronics. From the materials perspective, the available conducting polymers with self-healing properties remain limited. Response time and efficiency are two principal metrics when considering the healing properties. Improved performance with rapid recovery and high efficiency are always needed. Furthermore, long-term stability for healing function does face challenges, especially in the case of hydrogels with a high water content that is easily evaporated.
Additionally, due to a lack of electronic materials, research on self-healing electronic devices, or OECTs, is still restricted. The mismatching between layers makes it challenging to create fully healable OECTs with all self-healing layers. In spite of these challenges, a self-healing integrated electronic platform with multicomponents is expected.
Related Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Li Y, Zhou X, Sarkar B, Gagnon‐Lafrenais N, Cicoira F. 2022. Recent Progress on Self‐Healable Conducting Polymers. Advanced Materials. 34(24): https://doi.org/10.1002/adma.202108932
2.
Fu Y, Zhao J, Dong Y, Wang X. Dry Electrodes for Human Bioelectrical Signal Monitoring. Sensors. 20(13):3651. https://doi.org/10.3390/s20133651
3.
Bettucci O, Matrone GM, Santoro F. 2022. Conductive Polymer‐Based Bioelectronic Platforms toward Sustainable and Biointegrated Devices: A Journey from Skin to Brain across Human Body Interfaces. Adv Materials Technologies. 7(2): https://doi.org/10.1002/admt.202100293
4.
Malliaras G, McCulloch I. 2022. Introduction: Organic Bioelectronics. Chem. Rev. 122(4):4323-4324. https://doi.org/10.1021/acs.chemrev.2c00026
5.
Wang S, Urban MW. Self-healing polymers. Nat Rev Mater. 5(8):562-583. https://doi.org/10.1038/s41578-020-0202-4
6.
Roy N, Bruchmann B, Lehn J. DYNAMERS: dynamic polymers as self-healing materials. Chem. Soc. Rev. 44(11):3786-3807. https://doi.org/10.1039/c5cs00194c
7.
Shi H, Liu C, Jiang Q, Xu J. 2015. Effective Approaches to Improve the Electrical Conductivity of PEDOT:PSS: A Review. Adv Elect Materials. 1(4): https://doi.org/10.1002/aelm.201500017
8.
Zhang S, Kumar P, Nouas AS, Fontaine L, Tang H, Cicoira F. 2015. Solvent-induced changes in PEDOT:PSS films for organic electrochemical transistors. APL Mater. 3(1): https://doi.org/10.1063/1.4905154
9.
Zhang S, Cicoira F. 2017. Water‐Enabled Healing of Conducting Polymer Films. Advanced Materials. 29(40): https://doi.org/10.1002/adma.201703098
10.
Li Y, Zhang S, Hamad N, Kim K, Liu L, Lerond M, Cicoira F. 2020. Tailoring the Self‐Healing Properties of Conducting Polymer Films. Macromolecular Bioscience. 20(11): https://doi.org/10.1002/mabi.202000146
11.
Li Y, Li X, Zhang S, Liu L, Hamad N, Bobbara SR, Pasini D, Cicoira F. 2020. Autonomic Self‐Healing of PEDOT:PSS Achieved Via Polyethylene Glycol Addition. Adv Funct Materials. 30(30): https://doi.org/10.1002/adfm.202002853
12.
Oh JY, Kim S, Baik H, Jeong U. 2016. Conducting Polymer Dough for Deformable Electronics. Advanced Materials. 28(22):4455-4461. https://doi.org/10.1002/adma.201502947
13.
Huyer C, Modafferi D, Aminzare M, Ferraro J, Abdali Z, Roy S, Saldanha DJ, Wasim S, Alberti J, Feng S, et al. 2023. Fabrication of Curli Fiber-PEDOT:PSS Biomaterials with Tunable Self-Healing, Mechanical, and Electrical Properties. ACS Biomater. Sci. Eng. 9(5):2156-2169. https://doi.org/10.1021/acsbiomaterials.1c01180
14.
Zhou X, Rajeev A, Subramanian A, Li Y, Rossetti N, Natale G, Lodygensky GA, Cicoira F. 2022. Self-healing, stretchable, and highly adhesive hydrogels for epidermal patch electrodes. Acta Biomaterialia. 139296-306. https://doi.org/10.1016/j.actbio.2021.07.069
15.
Ma D, Wu X, Wang Y, Liao H, Wan P, Zhang L. 2019. Wearable, Antifreezing, and Healable Epidermal Sensor Assembled from Long-Lasting Moist Conductive Nanocomposite Organohydrogel. ACS Appl. Mater. Interfaces. 11(44):41701-41709. https://doi.org/10.1021/acsami.9b15412
16.
Wu J, Wu Z, Lu X, Han S, Yang B, Gui X, Tao K, Miao J, Liu C. 2019. Ultrastretchable and Stable Strain Sensors Based on Antifreezing and Self-Healing Ionic Organohydrogels for Human Motion Monitoring. ACS Appl. Mater. Interfaces. 11(9):9405-9414. https://doi.org/10.1021/acsami.8b20267
17.
Hu C, Zhang Y, Wang X, Xing L, Shi L, Ran R. 2018. Stable, Strain-Sensitive Conductive Hydrogel with Antifreezing Capability, Remoldability, and Reusability. ACS Appl. Mater. Interfaces. 10(50):44000-44010. https://doi.org/10.1021/acsami.8b15287
18.
Rong Q, Lei W, Chen L, Yin Y, Zhou J, Liu M. 2017. Anti‐freezing, Conductive Self‐healing Organohydrogels with Stable Strain‐Sensitivity at Subzero Temperatures. Angew Chem Int Ed. 56(45):14159-14163. https://doi.org/10.1002/anie.201708614
19.
Azimi M, Subramanian A, Fan J, Soavi F, Cicoira F. Electrical and mechanical stability of flexible, organic electrolyte-gated transistors based on iongel and hydrogels. J. Mater. Chem. C. 11(14):4623-4633. https://doi.org/10.1039/d3tc00410d
20.
Kim C, Azimi M, Fan J, Nagarajan H, Wang M, Cicoira F. All-printed and stretchable organic electrochemical transistors using a hydrogel electrolyte. Nanoscale. 15(7):3263-3272. https://doi.org/10.1039/d2nr06731e
21.
Azimi M, Kim C, Fan J, Cicoira F. Effect of ionic conductivity of electrolyte on printed planar and vertical organic electrochemical transistors. Faraday Discuss. 246540-555. https://doi.org/10.1039/d3fd00065f
22.
Rivnay J, Inal S, Salleo A, Owens RM, Berggren M, Malliaras GG. Organic electrochemical transistors. Nat Rev Mater. 3(2): https://doi.org/10.1038/natrevmats.2017.86
23.
Leleux P, Rivnay J, Lonjaret T, Badier J, Bénar C, Hervé T, Chauvel P, Malliaras GG. 2015. Organic Electrochemical Transistors for Clinical Applications. Adv Healthcare Materials. 4(1):142-147. https://doi.org/10.1002/adhm.201400356
24.
Nawaz A, Liu Q, Leong WL, Fairfull‐Smith KE, Sonar P. 2021. Organic Electrochemical Transistors for In Vivo Bioelectronics. Advanced Materials. 33(49): https://doi.org/10.1002/adma.202101874
25.
Bai L, Elósegui CG, Li W, Yu P, Fei J, Mao L. Biological Applications of Organic Electrochemical Transistors: Electrochemical Biosensors and Electrophysiology Recording. Front. Chem. 7 https://doi.org/10.3389/fchem.2019.00313
26.
Kang J, Tok JB, Bao Z. Self-healing soft electronics. Nat Electron. 2(4):144-150. https://doi.org/10.1038/s41928-019-0235-0
27.
Ko J, Wu X, Surendran A, Muhammad BT, Leong WL. 2020. Self-Healable Organic Electrochemical Transistor with High Transconductance, Fast Response, and Long-Term Stability. ACS Appl. Mater. Interfaces. 12(30):33979-33988. https://doi.org/10.1021/acsami.0c07913
28.
Zhang S, Chen Y, Liu H, Wang Z, Ling H, Wang C, Ni J, Çelebi‐Saltik B, Wang X, Meng X, et al. 2020. Room‐Temperature‐Formed PEDOT:PSS Hydrogels Enable Injectable, Soft, and Healable Organic Bioelectronics. Advanced Materials. 32(1): https://doi.org/10.1002/adma.201904752
29.
Su X, Wu X, Chen S, Nedumaran AM, Stephen M, Hou K, Czarny B, Leong WL. 2022. A Highly Conducting Polymer for Self‐Healable, Printable, and Stretchable Organic Electrochemical Transistor Arrays and Near Hysteresis‐Free Soft Tactile Sensors. Advanced Materials. 34(19): https://doi.org/10.1002/adma.202200682
30.
Han S, Yu S, Hu S, Chen H, Wu J, Liu C. A high endurance, temperature-resilient, and robust organic electrochemical transistor for neuromorphic circuits. J. Mater. Chem. C. 9(35):11801-11808. https://doi.org/10.1039/d1tc02215f