HPLC Analysis of Mepivacaine Enantiomers on Astec® CHIROBIOTIC® V2
Materials
analytical column
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
[A] methanol; [B] 20 mM ammonium acetate, pH 4.1; (95:5, A:B)
flow rate
1 mL/min
column temp.
20 °C
detector
UV, 220 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC