HPLC Analysis of Ibuprofen & Codeine Phosphate Tablet on Chromolith® High Resolution RP-18e
CONDITIONS
sample preparation
Weigh 20 tablet & crush. Tranfer equivalent to one tablet weight to a 100 ml volumetric flask. disperse in 70 ml methanol & sonicate for 20 min. Dilute upto the mark with methanol. Pipette out 5 ml of the above solution & dilute to 50 ml with mobile phase.
column
Chromolith High Resolution RP-18 endcapped, 100 x 4.6 m (1.52022.0001)
mobile phase
[A] 2.24 g of KH2PO4 & 5.4 g of Sodium Octane sulfonate in 1000 ml water containing 7 ml triethyl amine - adjust pH to 7.2 with ortho-phosphoric acid; [B] methanol; (70:30, A:B)
flow rate
1.0 mL/min
column temp.
40 °C
detector
214 nm cell: 10 μL
injection
10 μL
sample
200 ppm of Ibuprofen & 12.8 ppm of codeine phosphate in mobile phase.
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany