HPLC Analysis of Hydroxyzine Enantiomers on Astec® Cellulose DMP
![HPLC Analysis of Hydroxyzine Enantiomers on Astec® Cellulose DMP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/270/578/8d982072-a358-4985-9084-1a9954e76e1a/800/8d982072-a358-4985-9084-1a9954e76e1a.jpg)
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] heptane; [B] IPA; [C] DEA; (90:10:0.1, A:B:C)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
2 μL
sample
2 mg/mL in mobile phase