GC Analysis of FAMEs in Rapeseed Oil on SUPELCOWAX 10
Materials
analytical column
CONDITIONS
column
SUPELCOWAX 10, 30m × 0.53 mm I.D., 1.0 μm (25301-U)
oven
240 °C
inj. temp.
250 °C
detector
FID, 260 °C
carrier gas
helium, 5 mL/min (flow controlled)
injection
0.1 μL direct
sample
Rapeseed Oil Reference mix (O7756-1AMP)
Description
Legal Information
null