- ALKBH1-Mediated tRNA Demethylation Regulates Translation.
ALKBH1-Mediated tRNA Demethylation Regulates Translation.
tRNA is a central component of protein synthesis and the cell signaling network. One salient feature of tRNA is its heavily modified status, which can critically impact its function. Here, we show that mammalian ALKBH1 is a tRNA demethylase. It mediates the demethylation of N