- Selective inhibition of herpes simplex virus by 5-amino-2,5-dideoxy-5-iodouridine.
Selective inhibition of herpes simplex virus by 5-amino-2,5-dideoxy-5-iodouridine.
Journal of virology (1975-05-01)
Y C Cheng, B Goz, J P Neenan, D C Ward, W H Prusoff
PMID167186
ABSTRACT
5-Amino-2,5-dideoxy-5-iodouridine, a nel thymidine analogue, is a potent inhibitor of herpes simplex virus type 1 replication. In contrast to most other nucleoside analogues which possess antiviral activity, 5-amino-2,5-dideoxy-5-iodouridine exhibits little, if any, cellular toxicity. Preliminary evidence suggests that 5-amino-2,5-dideoxy-5-iodouridine selectively inhibits viral-specific DNA synthesis.