MilliporeSigma
  • Epigenetic and Transcriptional Alterations in Human Adipose Tissue of Polycystic Ovary Syndrome.
Epigenetic and Transcriptional Alterations in Human Adipose Tissue of Polycystic Ovary Syndrome.
Scientific reports (2016-03-16)
Milana Kokosar, Anna Benrick, Alexander Perfilyev, Romina Fornes, Emma Nilsson, Manuel Maliqueo, Carl Johan Behre, Antonina Sazonova, Claes Ohlsson, Charlotte Ling, Elisabet Stener-Victorin
ABSTRACT
Genetic and epigenetic factors may predispose women to polycystic ovary syndrome (PCOS), a common heritable disorder of unclear etiology. Here we investigated differences in genome-wide gene expression and DNA methylation in adipose tissue from 64 women with PCOS and 30 controls. In total, 1720 unique genes were differentially expressed (Q < 0.05). Six out of twenty selected genes with largest expression difference (CYP1B1, GPT), genes linked to PCOS (RAB5B) or type 2 diabetes (PPARG, SVEP1), and methylation (DMAP1) were replicated in a separate case-control study. In total, 63,213 sites (P < 0.05) and 440 sites (Q < 0.15) were differently methylated. Thirty differentially expressed genes had corresponding changes in 33 different DNA methylation sites. Moreover, a total number of 1913 pairs of differentially expressed "gene-CpG" probes were significantly correlated after correction for multiple testing and corresponded with 349 unique genes. In conclusion, we identified a large number of genes and pathways that are affected in adipose tissue from women with PCOS. We also identified specific DNA methylation pathways that may affect mRNA expression. Together, these novel findings show that women with PCOS have multiple transcriptional and epigenetic changes in adipose tissue that are relevant for development of the disease.