MilliporeSigma
  • Follicular fluid biomarkers for human in vitro fertilization outcome: Proof of principle.
Follicular fluid biomarkers for human in vitro fertilization outcome: Proof of principle.
Proteome science (2016-11-30)
Fang Chen, Carl Spiessens, Thomas D'Hooghe, Karen Peeraer, Sebastien Carpentier
ABSTRACT
Human follicular fluid (FF) is a unique biological fluid in which the oocyte develops in vivo, and presents an optimal source for non-invasive biochemical predictors. Oocyte quality directly influences the embryo development and hence, may be used as a predictor of embryo quality. Peptide profiling of FF and its potential use as a biomarker for oocyte quality has never been reported. This study screened FF for peptide biomarkers that predict the outcome of in vitro fertilization (IVF). Potential biomarkers were discovered by investigating 2 training datasets, consisting both of 17 samples and validating on an independent experiment containing 32 samples. Peptide profiles were acquired by nano-scale liquid chromatography coupled to tandem mass spectrometry (nano LC-MS/MS). From the training datasets 53 peptides were found as potential biomarker candidates, predicting the fertilization outcome of 24 out of the 32 validation samples blindly (81.3% sensitivity, 68.8% specificity, AUC = 0.86). Seven potential biomarker peptides were identified. They were derived from: insulin-like growth factor binding protein-5, alpha-2-antiplasmin, complement component 3, inter-alpha-trypsin inhibitor heavy chain H1, serum albumin, protein diaphanous homolog 1 and plastin-3. The MS-based comprehensive peptidomic approach carried out in this study, established a novel panel of potential biomarkers that present a promising predictive accuracy rate in fertilization outcome, and indicates FF as an interesting biomarker resource to improve IVF clinic routine.