Merck
  • Studies on angiotensin converting enzyme inhibitors. 4. Synthesis and angiotensin converting enzyme inhibitory activities of 3-acyl-1-alkyl-2-oxoimidazolidine-4-carboxylic acid derivatives.
Studies on angiotensin converting enzyme inhibitors. 4. Synthesis and angiotensin converting enzyme inhibitory activities of 3-acyl-1-alkyl-2-oxoimidazolidine-4-carboxylic acid derivatives.
Journal of medicinal chemistry (1989-02-01)
K Hayashi, K Nunami, J Kato, N Yoneda, M Kubo, T Ochiai, R Ishida
ABSTRACT
(4S)-1-Alkyl-3-[[N-(carboxyalkyl)amino]acyl]-2-oxoimidazolidine-4- carboxylic acid derivatives (3) were prepared by two methods. Their angiotensin converting enzyme (ACE) inhibitory activities and antihypertensive effects were evaluated, and the structure-activity relationships were discussed. The dicarboxylic acids 3a-n possessing S,S,S configuration showed potent in vitro ACE inhibitory activities with IC50 values of 1.1 X 10(-8)-1.5 X 10(-9) M. The most potent compound in this series, monoester 3p, had an ID50 value of 0.24 mg/kg, po for inhibition of angiotensin I induced pressor response in normotensive rats and produced a dose-dependent decrease in systolic blood pressure of spontaneously hypertensive rats (SHRs) at doses of 1-10 mg/kg, po.
Sigma-Aldrich
(S)-(−)-1-Z-2-Oxo-5-imidazolidinecarboxylic acid, 98%
Pricing and availability is not currently available.