Merck
  • TLC-Direct Bioautography as a Bioassay Guided Method for Investigation of Antibacterial Compounds in Hypericum perforatum L.
TLC-Direct Bioautography as a Bioassay Guided Method for Investigation of Antibacterial Compounds in Hypericum perforatum L.
Journal of AOAC International (2015-08-14)
Wioleta Jesionek, Ágnes M Móricz, Ágnes Alberti, Péter G Ott, Béla Kocsis, Györgyi Horváth, Irena M Choma
ABSTRACT
Fast high-throughput TLC-direct bioautography (DB) is an effect-directed analysis method that enables searching for biologically active (e.g., antimicrobial) substances in complex mixtures like plant extracts. The principle of the method is that separation and detection of biological properties of given mixture components is performed directly on a TLC plate. In searching for antibacterial activity, the developed plate is immersed in a bacterial broth, and bacteria grow directly on its layer during a proper incubation time. Inhibition zones are formed in places where antimicrobial components are located. The active compounds can be further identified using spectroscopic techniques. The aim of our study was investigation of plant components of Hypericum perforatum L. tincture by TLC-DB using nine bacterial strains: Micrococcus luteus, Bacillus subtilis, Escherichia coli, Staphylococcus aureus, methicillin-resistant S. aureus, S. epidermidis, Pseudomonas syringae pv. maculicola, Xanthomonas campestris pv. vesicatoria, and Aliivibrio fischeri. Compounds showing the widest range of antimicrobial activity were isolated using semipreparative TLC and identified as apigenin, 3,8'-biapigenin, quercetin, kaempferol, and linolenic acid by TLC, HPLC-diode array detection, and HPLC/MS/MS techniques.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
p-Anisaldehyde, 98%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
p-Anisaldehyde, natural, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder