Merck
  • Structural and kinetic analysis of the substrate specificity of human fibroblast activation protein alpha.
Structural and kinetic analysis of the substrate specificity of human fibroblast activation protein alpha.
The Journal of biological chemistry (2005-04-06)
Kathleen Aertgeerts, Irena Levin, Lihong Shi, Gyorgy P Snell, Andy Jennings, G Sridhar Prasad, Yanming Zhang, Michelle L Kraus, Shant Salakian, Vandana Sridhar, Robert Wijnands, Mike G Tennant
ABSTRACT
Fibroblast activation protein alpha (FAPalpha) is highly expressed in epithelial cancers and has been implicated in extracellular matrix remodeling, tumor growth, and metastasis. We present the first high resolution structure for the apoenzyme as well as kinetic data toward small dipeptide substrates. FAPalpha exhibits a dipeptidyl peptidase IV (DPPIV)-like fold, featuring an alpha/beta-hydrolase domain and an eight-bladed beta-propeller domain. Known DPPIV dipeptides are cleaved by FAPalpha with an approximately 100-fold decrease in catalytic efficiency compared with DPPIV. Moreover, FAPalpha, but not DPPIV, possesses endopeptidase activity toward N-terminal benzyloxycarbonyl (Z)-blocked peptides. Comparison of the crystal structures of FAPalpha and DPPIV revealed one major difference in the vicinity of the Glu motif (Glu(203)-Glu(204) for FAPalpha; Glu(205)-Glu(206) for DPPIV) within the active site of the enzyme. Ala(657) in FAPalpha, instead of Asp(663) as in DP-PIV, reduces the acidity in this pocket, and this change could explain the lower affinity for N-terminal amines by FAPalpha. This hypothesis was tested by kinetic analysis of the mutant FAPalpha/A657D, which shows on average an approximately 60-fold increase in the catalytic efficiency, as measured by k(cat)/K(m), for the cleavage of dipeptide substrates. Furthermore, the catalytic efficiency of the mutant is reduced by approximately 350-fold for cleavage of Z-Gly-Pro-7-amino-4-methylcoumarin. Our data provide a clear understanding of the molecular determinants responsible for the substrate specificity and endopeptidase activity of FAPalpha.
Sigma-Aldrich
Fibroblast Activation Protein α human, recombinant, expressed in Sf21 cells, ≥95% (SDS-PAGE)