Merck
  • Dual blockade of the renin-angiotensin-aldosterone system with high-dose angiotensin-converting enzyme inhibitor for nephroprotection: an open, controlled, randomized study.
Dual blockade of the renin-angiotensin-aldosterone system with high-dose angiotensin-converting enzyme inhibitor for nephroprotection: an open, controlled, randomized study.
Scandinavian journal of urology and nephrology (2009-02-21)
Leszek Tylicki, Marcin Renke, Przemysław Rutkowski, Wojciech Larczyński, Ewa Aleksandrowicz, Wiesława Lysiak-Szydlowska, Bolesław Rutkowski
ABSTRACT
Despite the proven effectiveness of combination therapy with an angiotensin I-converting enzyme inhibitor (ACEI) and angiotensin II-receptor blockers (ARBs) for the prevention and treatment of kidney disease, it has not proved possible to inhibit the progress of chronic nephropathies completely. To improve renal outcome one may consider using increased dosages of ACEI above those usually recommended for hypertension. A randomized, open, controlled study was conducted to evaluate the influence of two combination therapies on proteinuria, markers of tubular injury and renal fibrosis. A total of 18 patients with a creatinine level of 109+/-36 micromol/l and proteinuria of 0.97+/-0.76 g/24 h were enrolled in the study. In the 8-week run-in period, an ACEI (cilazapril 5 mg once-daily) and an ARB (telmisartan 80 mg once-daily) were administered to achieve the target blood pressure of < or = 130/80 mmHg. Next, the patients were randomly assigned to either an increased dose of cilazapril (10 mg) or the previous dose (5 mg) in two active-treatment periods, each lasting 8 weeks. A significant increase in renin activity was observed after administration of cilazapril 10 mg (6.46+/-1.12 vs 4.67+/-0.7 ng/ml/h; p=0.028). Proteinuria, urine excretion of N-acetyl-beta-D-glucosaminidase, and alpha1-microglobulin and amino-terminal propeptide of type III procollagen were unchanged. An increased dosage of cilazapril (twice the maximum recommended dose) in addition to combination therapy with telmisartan was associated with increased blockade of the renin-angiotensin-aldosterone system, with no additional effect on proteinuria, markers of tubular injury or renal fibrosis.
Product Description
Cilazapril impurity D, European Pharmacopoeia (EP) Reference Standard
Cilazapril, European Pharmacopoeia (EP) Reference Standard