Merck
  • Protective effects of calcium gluconate on osteoarthritis induced by anterior cruciate ligament transection and partial medial meniscectomy in Sprague-Dawley rats.
Protective effects of calcium gluconate on osteoarthritis induced by anterior cruciate ligament transection and partial medial meniscectomy in Sprague-Dawley rats.
Journal of orthopaedic surgery and research (2014-03-08)
Su-Jin Kang, Joo-Wan Kim, Ki-Young Kim, Sae-Kwang Ku, Young-Joon Lee
ABSTRACT
This study aimed to determine whether calcium gluconate exerts protective effects on osteoarthritis (OA) induced by anterior cruciate ligament (ACL) transection and partial medial meniscectomy. Calcium gluconate was administered by mouth daily for 84 days to male ACL transected and partial medial meniscectomized Sprague-Dawley rats 1 week after operation. Eighty-four days of treatment with 50 mg/kg calcium gluconate led to a lower degree of articular stiffness and cartilage damage compared to the OA control, possibly through inhibition of overexpressed cyclooxygenase (COX)-2 and related chondrocyte apoptosis. Similar favorable effects on stiffness and cartilage were detected in calcium gluconate-administered rats. Additionally, calcium gluconate increased 5-bromo-2'-deoxyuridine (BrdU) uptake based on observation of BrdU-immunoreactive cells on both the femur and tibia articular surface cartilages 84 days after intra-joint treatment with calcium gluconate. Taken together, our results demonstrate that calcium gluconate has a protective effect against OA through inhibition of COX-2 and related chondrocyte apoptosis.