- 2-Pyridinyl beta-ketones as new ligands for room-temperature CuI-catalysed C-N coupling reactions.
2-Pyridinyl beta-ketones as new ligands for room-temperature CuI-catalysed C-N coupling reactions.
Chemical communications (Cambridge, England) (2009-03-26)
Deping Wang, Ke Ding
PMID19319436
ABSTRACT
2-Pyridinyl beta-ketones were identified as new efficient ligands for CuI-catalysed N-arylation of aliphatic amines at room temperature with great selectivity and substrate scope tolerance.