- Influence of chromium on some physiological variables of Anabaena doliolum: interaction with metabolic inhibitors.
Influence of chromium on some physiological variables of Anabaena doliolum: interaction with metabolic inhibitors.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (1992-01-01)
L C Rai, S K Dubey, N Mallick
PMID1392466
ABSTRACT
The impact of 2,4-dinitrophenol and chlorophenyl dimethylurea on ATP content, carbon fixation, O2 evolution, nitrogenase activity and Cr uptake of Anabaena doliolum has been studied. 2,4-Dinitrophenol has been found to be more toxic than chlorophenyldimethylurea for all these processes. However, when Cr toxicity to above variables was assessed in their presence the interaction was less than additive. An initial (10-15 min) concentration-dependent rapid Cr uptake, followed by a slow one, indicates a biphasic uptake. A significant inhibition of Cr uptake in the presence of both these metabolic inhibitors suggests the involvement of metabolic processes in Cr uptake.