MilliporeSigma
  • Proposed model for in vitro interaction between fenitrothion and DNA, by using competitive fluorescence, (31)P NMR, (1)H NMR, FT-IR, CD and molecular modeling.
Proposed model for in vitro interaction between fenitrothion and DNA, by using competitive fluorescence, (31)P NMR, (1)H NMR, FT-IR, CD and molecular modeling.
Toxicology in vitro : an international journal published in association with BIBRA (2012-11-17)
Farhad Ahmadi, Batool Jafari, Mehdi Rahimi-Nasrabadi, Sahar Ghasemi, Kumars Ghanbari
ABSTRACT
In this work we proposed a model for in vitro interaction of fenitrothion (FEN) with calf thymus-DNA by combination of multispectroscopic and two dimensional molecular modeling (ONIOM) methods. The circular dichroism results showed that FEN changes the conformation of B-DNA and caused some changes to C-DNA form. The FT-IR results confirmed a partial intercalation between FEN and edges of all base pairs. The competitive fluorescence, using methylene blue as fluorescence probe, in the presence of increasing amounts of FEN, revealed that FEN is able to release the non-intercalated methylene blue from the DNA. The weak chemical shift and peak broadening of (1)H NMR spectrum of FEN in the presence of DNA confirmed a non-intercalation mode. The (31)P NMR showed that FEN interacts more with DNA via its -NO2 moiety. The ONIOM, based on the hybridization of QM/MM (DFT, 6.31++G (d,p)/UFF) methodology, was also performed by Gaussian 2003 package. The results revealed that the interaction is base sequence dependent, and FEN interacts more with AT base sequences.
Product Description
Supelco
Fenitrothion, PESTANAL®, analytical standard
Pricing and availability is not currently available.