Merck
Flumethasone-induced insulin resistance in calves.
Zentralblatt fur Veterinarmedizin. Reihe A (1998-10-30)
K Sternbauer, J Luthman, S O Jacobsson
ABSTRACT
The impact of one single therapeutic dose of flumethasone (0.5 mg/100 kg b.w.) on insulin sensitivity was studied in calves. Hyperinsulinemic, euglycemic clamp tests were performed before and after flumethasone treatment. At 24 h after treatment, insulin-dependent glucose utilization was reduced by 74% (P < 0.005). No significant changes occurred 72 h post-treatment.
Sigma-Aldrich
Flumethasone
Pricing and availability is not currently available.