Merck
  • Clinical and immunohistopathological aspects of venous ulcers treatment by Low-Intensity Pulsed Ultrasound (LIPUS).
Clinical and immunohistopathological aspects of venous ulcers treatment by Low-Intensity Pulsed Ultrasound (LIPUS).
Ultrasonics (2013-01-09)
Luísiane de Ávila Santana, José Marcos Alves, Thiago Antônio Moretti Andrade, João Kazuyuki Kajiwara, Sérgio Britto Garcia, Fernanda Guzzo Gomes, Marco Andrey Cipriani Frade
ABSTRACT
The immunological mechanisms that are triggered by Low-Intensity Pulsed Ultrasound (LIPUS) in wound healing are unknown. In the present study, experimental groups were used to assess the treatment of chronic venous ulcers with 30mW/cm(2) SATA peripheral LIPUS three times per week compared to a daily treatment of 1% silver sulfadiazine (SDZ). The ulcers of the SDZ group (n=7) (G1) and LIPUS group (n=9) (G2) were photographed five times three months, and the images were analyzed using ImageJ software to quantify the total area (S), fibrin/sphacel area (yellow) and granulation area (red). The healing process was evaluated by the wound healing rate (WHR), granulation tissue rate (GTR) and fibrin/sphacel tissue rate (FTR). The ulcers were biopsied on days 1 and 45 and stained for collagen fiber quantification (picrosirius) and CD68(+) protein and VEGF (vascular endothelial growth factor) expression using HRP-streptavidin (horseradish peroxidase-streptavidin). On day 90, G2 had a mean 41% decrease in the ulcer area, while no decrease was observed in G1 (p<0.05). An increased tendency toward positive labeling of collagen fibers and VEGF (p>0.05) was observed in G2 compared to G1, and the number of CD68(+) cells was greater in G2 than in G1 (p<0.05). LIPUS presents superior activity compared to SDZ in stimulating the inflammatory and proliferative (angiogenesis and collagenesis, respectively) phases of chronic venous wound healing.
Sigma-Aldrich
Silver(I) sulfadiazine, 98%