Merck
  • Determination of the Alternaria mycotoxin tenuazonic acid in cereals by high-performance liquid chromatography-electrospray ionization ion-trap multistage mass spectrometry after derivatization with 2,4-dinitrophenylhydrazine.
Determination of the Alternaria mycotoxin tenuazonic acid in cereals by high-performance liquid chromatography-electrospray ionization ion-trap multistage mass spectrometry after derivatization with 2,4-dinitrophenylhydrazine.
Journal of chromatography. A (2009-04-14)
David Siegel, Tatjana Rasenko, Matthias Koch, Irene Nehls
ABSTRACT
Tenuazonic acid (TA) is a major Alternaria mycotoxin. In the present work a novel approach for the detection of TA in cereals by liquid chromatography-ion-trap multistage mass spectrometry after derivatization with 2,4-dinitrophenylhydrazine is described. The product of the derivatization reaction and its major MS(2) fragments were characterised by Fourier transform-ion cyclotron resonance tandem mass spectrometry. Without preconcentration, the established method features a limit of detection of 10 microg/kg using 2g of sample in a rapid workup procedure. Accuracy, precision and linearity were evaluated in the working range of 50-5000 microg/kg. TA was detected in 13 and quantified in 3 out of 27 cereal samples obtained from a local supermarket, the average content being 49 microg/kg (highest incidence: 851+/-41 microg/kg).
Sigma-Aldrich
Tenuazonic acid copper salt from Alternaria alternata