Merck
  • Catalysis of regioselective reduction of NAD+ by ruthenium(II) arene complexes under biologically relevant conditions.
Catalysis of regioselective reduction of NAD+ by ruthenium(II) arene complexes under biologically relevant conditions.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2006-04-11)
Yaw Kai Yan, Michael Melchart, Abraha Habtemariam, Anna F A Peacock, Peter J Sadler
ABSTRACT
Ruthenium(II) arene anticancer complexes [(eta6-arene)Ru(en)Cl]PF6 (arene is hexamethylbenzene, p-cymene, indan; en is ethylenediamine) can catalyse regioselective reduction of NAD+ by formate in water to form 1,4-NADH, at pD 7.2, 37 degrees C, and in the presence of air. The catalytic activity is markedly dependent on the arene, with the hexamethylbenzene (hmb) complex showing the highest activity. For [(eta 6-hmb)Ru(en)Cl]PF6, the rate of reaction is independent of NAD+ concentration and shows saturation kinetics with respect to formate concentration. A Km value of 58 mM and a turnover frequency at saturation of 1.46 h(-1) were observed. Removal of chloride and performing the reaction under argon led to higher reaction rates. Lung cancer cells (A549) were found to be remarkably tolerant to formate even at millimolar concentrations. The possibility of using ruthenium arene complexes coadministered with formate as catalytic drugs is discussed.
Sigma-Aldrich
Hexamethylbenzene, 99%