Merck
  • Leukocyte-endothelial cell interaction is necessary for photodynamic therapy induced vascular permeabilization.
Leukocyte-endothelial cell interaction is necessary for photodynamic therapy induced vascular permeabilization.
Lasers in surgery and medicine (2011-11-08)
Elodie Debefve, François Mithieux, Jean Y Perentes, Yabo Wang, Cai Cheng, Stephan C Schaefer, Christiane Ruffieux, Jean-Pierre Ballini, Michel Gonzalez, Hubert van den Bergh, Hans-Beat Ris, Hans-Anton Lehr, Thorsten Krueger
ABSTRACT
Photodynamic therapy (PDT) affects vascular barrier function and thus increases vessel permeability. This phenomenon may be exploited to facilitate targeted drug delivery and may lead to a new clinical application of photodynamic therapy. Here, we investigate the role of leukocyte recruitment for PDT-induced vascular permeabilization. Fluorescein isothiocyanate dextran (FITC-D, 2,000 kDa) was injected intravenously 120 minutes after focal PDT on striated muscle in nude mice bearing dorsal skinfold chambers (Visudyne® 800 µg/kg, fluence rate 300 mW/cm2 , light dose of 200 J/cm2). Leukocyte interaction with endothelial cells was inhibited by antibodies functionally blocking adhesion molecules ("MABS-PDT" group, n = 5); control animals had PDT but no antibody injection (group "PDT", n = 7). By intravital microscopy, we monitored leukocyte rolling and sticking in real-time before, 90 and 180 minutes after PDT. The extravasation of FITC-D from striated muscle vessels into the interstitial space was determined in vivo during 45 minutes to assess treatment-induced alterations of vascular permeability. PDT significantly increased the recruitment of leukocytes and enhanced the leakage of FITC-D. Neutralization of adhesion molecules before PDT suppressed the rolling of leukocytes along the venular endothelium and significantly reduced the extravasation of FITC-D as compared to control animals (156 ± 27 vs. 11 ± 2 (mean ± SEM, number of WBC/30 seconds mm vessel circumference; P < 0.05) at 90 minutes after PDT and 194 ± 21 vs. 14 ± 4 at 180 minutes after PDT). In contrast, leukocyte sticking was not downregulated by the antibody treatment. Leukocyte recruitment plays an essential role in the permeability-enhancing effect of PDT.
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 2,000,000