Merck
  • Replacing Commercial 6-Phosphofructokinase in an Online Pyrophosphate Detection Assay.
Replacing Commercial 6-Phosphofructokinase in an Online Pyrophosphate Detection Assay.
Chembiochem : a European journal of chemical biology (2022-07-26)
Daniel L Machell, Mathias H Hansen, Max J Cryle
ABSTRACT
Detection of pyrophosphate is important in quantifying enzyme activity, particularly adenylation domain activity during non-ribosomal peptide synthesis. The previous development of an enzyme coupled PPi /NADH assay allowed the measurement of such activity in an online fashion using commercially available components. Now, with a key enzyme - 6-phosphofructokinase - no longer available, we have screened and identified viable replacement enzymes that can be expressed in high yield and that are far superior in activity to the now discontinued commercial product. This will support the ability of groups to continue to use this established online assay for pyrophosphate detection.