Merck
  • Preparation and Detection of Glycan-Binding Activity of Influenza Virus.
Preparation and Detection of Glycan-Binding Activity of Influenza Virus.
Methods in molecular biology (Clifton, N.J.) (2020-04-20)
Shin-Ichi Nakakita, Nongluk Sriwilaijaroen, Yasuo Suzuki, Jun Hirabayashi
ABSTRACT
We describe a method to detect influenza virus using an evanescent-field-activated fluorescence scanner type glycan array and ELISA system. Neoglycoprotein was prepared by combination of organic chemistry and biomaterial preparation. These ligands were spotted on a glass plate or plastic well to make a glycan array and ELISA plate. We detected cultured influenza virus using glycan array and ELISA. Then, we showed that the neoglycoprotein binds to Cy3-labeled hemagglutinins (H1 and H5), a NeuAcα2,6LacNAc or NeuAcα2,3LacNAc recognized protein, as detected.