Merck
  • Coresistance to isoniazid and ethionamide maps to mycothiol biosynthetic genes in Mycobacterium bovis.
Coresistance to isoniazid and ethionamide maps to mycothiol biosynthetic genes in Mycobacterium bovis.
Antimicrobial agents and chemotherapy (2011-06-29)
Catherine Vilchèze, Yossef Av-Gay, S Whitney Barnes, Michelle H Larsen, John R Walker, Richard J Glynne, William R Jacobs
ABSTRACT
A search to identify new mechanisms of isoniazid resistance in Mycobacterium bovis led to the isolation of mutants defective in mycothiol biosynthesis due to mutations in genes coding for the glycosyltransferase (mshA) or the cysteine ligase (mshC). These mutants showed low-level resistance to isoniazid but were highly resistant to ethionamide. This study further illustrates that mutations in mycothiol biosynthesis genes may contribute to isoniazid or ethionamide resistance across mycobacterial species.
Sigma-Aldrich
Ethionamide