Merck
  • An immune response characterizes early Alzheimer's disease pathology and subjective cognitive impairment in hydrocephalus biopsies.
An immune response characterizes early Alzheimer's disease pathology and subjective cognitive impairment in hydrocephalus biopsies.
Nature communications (2021-09-29)
Wenrui Huang, Anne Marie Bartosch, Harrison Xiao, Suvrajit Maji, Elliot H H Youth, Xena Flowers, Sandra Leskinen, Zeljko Tomljanovic, Gail Iodice, Deborah Boyett, Eleonora Spinazzi, Vilas Menon, Robert A McGovern, Guy M McKhann, Andrew F Teich
ABSTRACT
Early Alzheimer's disease (AD) pathology can be found in cortical biopsies taken during shunt placement for Normal Pressure Hydrocephalus. This represents an opportunity to study early AD pathology in living patients. Here we report RNA-seq data on 106 cortical biopsies from this patient population. A restricted set of genes correlate with AD pathology in these biopsies, and co-expression network analysis demonstrates an evolution from microglial homeostasis to a disease-associated microglial phenotype in conjunction with increasing AD pathologic burden, along with a subset of additional astrocytic and neuronal genes that accompany these changes. Further analysis demonstrates that these correlations are driven by patients that report mild cognitive symptoms, despite similar levels of biopsy β-amyloid and tau pathology in comparison to patients who report no cognitive symptoms. Taken together, these findings highlight a restricted set of microglial and non-microglial genes that correlate with early AD pathology in the setting of subjective cognitive decline.
Sigma-Aldrich
Bovine Serum Albumin solution, 30%±2% in 0.85% sodium chloride, aseptically filled