Merck
  • MLL3 suppresses tumorigenesis through regulating TNS3 enhancer activity.
MLL3 suppresses tumorigenesis through regulating TNS3 enhancer activity.
Cell death & disease (2021-04-08)
Jun-Yi Zheng, Chen-Yu Wang, Chuan Gao, Qiong Xiao, Cheng-Wei Huang, Min Wu, Lian-Yun Li
ABSTRACT
MLL3 is a histone H3K4 methyltransferase that is frequently mutated in cancer, but the underlying molecular mechanisms remain elusive. Here, we found that MLL3 depletion by CRISPR/sgRNA significantly enhanced cell migration, but did not elevate the proliferation rate of cancer cells. Through RNA-Seq and ChIP-Seq approaches, we identified TNS3 as the potential target gene for MLL3. MLL3 depletion caused downregulation of H3K4me1 and H3K27ac on an enhancer ~ 7 kb ahead of TNS3. 3C assay indicated the identified enhancer interacts with TNS3 promoter and repression of enhancer activity by dCas9-KRAB system impaired TNS3 expression. Exogenous expression of TNS3 in MLL3 deficient cells completely blocked the enhanced cell migration phenotype. Taken together, our study revealed a novel mechanism for MLL3 in suppressing cancer, which may provide novel targets for diagnosis or drug development.