Merck
  • Reconstitution of two isoforms of the human interleukin-11 receptor and comparison of their functional properties.
Reconstitution of two isoforms of the human interleukin-11 receptor and comparison of their functional properties.
FEBS letters (1997-04-28)
B Lebeau, F A Montero Julian, J Wijdenes, G Müller-Newen, H Dahmen, M Chérel, P C Heinrich, H Brailly, M M Hallet, A Godard, S Minvielle, Y Jacques
ABSTRACT
Long-term stable Ba/F3 transfectants (B13R alpha1 and B13R alpha2) expressing two isoforms of the human IL-IIR alpha receptor (alpha1 full length or alpha2 lacking the cytoplasmic domain) in combination with human gp130 were established. IL-11R alpha1 and IL-11R alpha2 were each expressed and detected as three bands upon Western blot analysis, with apparent molecular masses in agreement with those of the polypeptide backbone (47 and 44 kDa, respectively) with no, one or two N-linked sugars. B13R alpha1 and B13R alpha2 bound IL-11-thioredoxin with similar efficiencies and proliferated with superimposable dose-response curves to IL-11, demonstrating that the intracellular domain of IL-11R alpha has no significant contribution on ligand binding and signaling. Analysis of a set of anti-human gp130 mAbs confirmed the similar responsiveness of B13R alpha1 and B13R alpha2 transfectants.