Merck
  • The prognostic significance of ALDH1A1 expression in early invasive breast cancer.
The prognostic significance of ALDH1A1 expression in early invasive breast cancer.
Histopathology (2020-05-06)
Maryam Althobiti, Rokaya El Ansari, Mohammed Aleskandarany, Chitra Joseph, Michael S Toss, Andrew R Green, Emad A Rakha
ABSTRACT
Aldehyde dehydrogenase family 1 member A1 (ALDH1A1) is reportedly a key ALDH isozyme linked to the cancer stem cells (CSC) of many solid tumours, where it is involved in self-renewal, differentiation and self-protection. In this study, the prognostic significance of ALDH1A1 expression in early invasive breast cancer (BC) and its role as a BC stem cell (BCSC) were evaluated. ALDH1A1 expression was assessed, using immunohistochemistry and tissue microarrays, in a large well-characterised BC cohort. ALDH1A1 mRNA expression was also assessed at transcriptomic levels, utilising data from the Molecular Taxonomy of Breast Cancer International Consortium. The associations of ALDH1A1 with clinicopathological parameters, other stem cell markers and patient outcomes were determined. ALDH1A1 was expressed in 71% of BC cases at both the protein and mRNA levels. High ALDH1A1 expression was associated with poor prognostic features, including high grade, poor Nottingham Prognostic Index (NPI), lymph node metastasis and highly proliferative ER+ (luminal B) and triple-negative (TNBC) subtypes. ALDH1A1 expression was positively correlated with the expression of CD44, CD24, TWIST, SOX9, EPCAM and CD133. The high immunoexpression of ALDH1A1 was significantly associated with poor BC-specific survival (P < 0.001), and specifically in the luminal B and TNBC subtypes (P = 0.042 and P = 0.003, respectively). The immunoexpression of ALDH1A1 was an independent predictor of poor prognosis (P = 0.015). ALDH1A1, as assessed using immunohistochemistry, seems to act as a BCSC marker associated not only with other BCSC markers but also with poor prognostic characteristics and poor outcomes, particularly in the luminal B and TNBC subtypes.
Sigma-Aldrich
Anti-ALDH1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution