Merck
  • Curcumin induces apoptosis in human breast cancer cells through p53-dependent Bax induction.
Curcumin induces apoptosis in human breast cancer cells through p53-dependent Bax induction.
FEBS letters (2002-02-20)
Tathagata Choudhuri, Suman Pal, Munna L Agwarwal, Tanya Das, Gaurisankar Sa
ABSTRACT
The aim of this study was to determine the mechanisms of curcumin-induced human breast cancer cell apoptosis. From quantitative image analysis data showing an increase in the percentage of cells with a sub-G0/G1 DNA content, we demonstrated curcumin-induced apoptosis in the breast cancer cell line MCF-7, in which expression of wild-type p53 could be induced. Apoptosis was accompanied by an increase in p53 level as well as its DNA-binding activity followed by Bax expression at the protein level. Further experiments using p53-null MDAH041 cell as well as low and high p53-expressing TR9-7 cell, in which p53 expression is under tight control of tetracycline, established that curcumin induced apoptosis in tumor cells via a p53-dependent pathway in which Bax is the downstream effector of p53. This property of curcumin suggests that this molecule could have a possible therapeutic potential in breast cancer patients.