- Effects of Notch signalling on the expression of SEMA3C, HMGA2, CXCL14, CXCR7, and CCL20 in breast cancer.
Effects of Notch signalling on the expression of SEMA3C, HMGA2, CXCL14, CXCR7, and CCL20 in breast cancer.
Metastasis is the main reason for death in breast cancer. Understanding the molecular players in metastasis is crucial for diagnostic and therapeutic purposes. Notch signalling plays an oncogenic role in breast tumorigenesis and is involved in metastasis. Downstream mediators of Notch signalling in prometastatic processes are not yet fully discovered. Here we aimed to investigate whether Notch signalling regulates the expression of SEMA3C, HMGA2, CXCL14, CXCR7, and CCL20, which are involved in prometastatic processes, in breast cell lines. To this end, expression of the selected genes was analysed following Notch activation by overexpression of the Notch1 intracellular domain in the normal breast epithelial cell line MCF10A, and inhibition by silencing of the Notch transcriptional mediator RBPjκ in the breast cancer cell line MDA MB 231. SEMA3C and HMGA2 mRNA were decreased, while CXCL14 and CXCR7 mRNA were increased significantly in response to Notch activation in MCF10A cells. Notch inhibition in MDA MB 231 cells significantly decreased HMGA2 and CCL20 mRNA. Protein levels were not significantly altered by Notch modulation. In conclusion, we showed that Notch signalling regulates expression of SEMA3C, CXCL14, CCL20, CXCR7, and HMGA2, which are prominent candidate genes that might function downstream of Notch to induce prometastatic processes.