Merck
  • Peroxynitrite targets the epidermal growth factor receptor, Raf-1, and MEK independently to activate MAPK.
Peroxynitrite targets the epidermal growth factor receptor, Raf-1, and MEK independently to activate MAPK.
The Journal of biological chemistry (2000-05-10)
P Zhang, Y Z Wang, E Kagan, J C Bonner
ABSTRACT
Activation of ERK-1 and -2 by H(2)O(2) in a variety of cell types requires epidermal growth factor receptor (EGFR) phosphorylation. In this study, we investigated the activation of ERK by ONOO(-) in cultured rat lung myofibroblasts. Western blot analysis using anti-phospho-ERK antibodies along with an ERK kinase assay using the phosphorylated heat- and acid-stable protein (PHAS-1) substrate demonstrated that ERK activation peaked within 15 min after ONOO(-) treatment and was maximally activated with 100 micrometer ONOO(-). Activation of ERK by ONOO(-) and H(2)O(2) was blocked by the antioxidant N-acetyl-l-cysteine. Catalase blocked ERK activation by H(2)O(2), but not by ONOO(-), demonstrating that the effect of ONOO(-) was not due to the generation of H(2)O(2). Both H(2)O(2) and ONOO(-) induced phosphorylation of EGFR in Western blot experiments using an anti-phospho-EGFR antibody. However, the EGFR tyrosine kinase inhibitor AG1478 abolished ERK activation by H(2)O(2), but not by ONOO(-). Both H(2)O(2) and ONOO(-) activated Raf-1. However, the Raf inhibitor forskolin blocked ERK activation by H(2)O(2), but not by ONOO(-). The MEK inhibitor PD98059 inhibited ERK activation by both H(2)O(2) and ONOO(-). Moreover, ONOO(-) or H(2)O(2) caused a cytotoxic response of myofibroblasts that was prevented by preincubation with PD98059. In a cell-free kinase assay, ONOO(-) (but not H(2)O(2)) induced autophosphorylation and nitration of a glutathione S-transferase-MEK-1 fusion protein. Collectively, these data indicate that ONOO(-) activates EGFR and Raf-1, but these signaling intermediates are not required for ONOO(-)-induced ERK activation. However, MEK-1 activation is required for ONOO(-)-induced ERK activation in myofibroblasts. In contrast, H(2)O(2)-induced ERK activation is dependent on EGFR activation, which then leads to downstream Raf-1 and MEK-1 activation.
Sigma-Aldrich
MEK1 Protein, inactive, 50 g, Unactive, N-terminal GST & C-terminal His6-tagged, recombinant full-length human MEK1, for use in Kinase Assays.