![Voriconazole Related Compound F United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/547/143/cb83332e-dec2-4c41-9573-413077ad5515/640/cb83332e-dec2-4c41-9573-413077ad5515.png)
Voriconazole Related Compound F
![Voriconazole Related Compound F United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/547/143/cb83332e-dec2-4c41-9573-413077ad5515/640/cb83332e-dec2-4c41-9573-413077ad5515.png)
Synonym(s):
(±)-Camphor-10-sulfonic acid, (±)-β-Camphorsulfonic acid
Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing