Skip to Content
Merck
trans-1,4-Diaminocyclohexane ≥98.0% (GC)

trans-1,4-Diaminocyclohexane

trans-1,4-Diaminocyclohexane ≥98.0% (GC)
Synonym(s):
trans-1,4-Cyclohexanediamine
Linear Formula:
C6H10(NH2)2
CAS Number:
Molecular Weight:
114.19

Products

Shipping
Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing