![Tolazoline hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/424/930/5dc7d25f-c7f6-47db-a65b-66555b470b79/640/5dc7d25f-c7f6-47db-a65b-66555b470b79.png)
Tolazoline hydrochloride
![Tolazoline hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/424/930/5dc7d25f-c7f6-47db-a65b-66555b470b79/640/5dc7d25f-c7f6-47db-a65b-66555b470b79.png)
Synonym(s):
2-Benzyl-2-imidazoline, 2-Benzyl-2-imidazoline hydrochloride
Empirical Formula (Hill Notation):
C10H12N2 · HCl
CAS Number:
Molecular Weight:
196.68
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing