Skip to Content
KR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium(IV) chloride
Synonym(s):
TTC, Titanium tetrachloride
Linear Formula:
TiCl
4
CAS Number:
7550-45-0
Molecular Weight:
189.68
EC Number:
231-441-9
Products
Product Category
acids (1)
salts (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
208566
Titanium(IV) chloride,
ReagentPlus
®
, 99.9% trace metals basis
Expand
View Pricing
254312
Titanium(IV) chloride,
≥99.995% trace metals basis
Expand
View Pricing