![Sodium deoxycholate monohydrate BioUltra, ≥99.0% (NT)](/deepweb/assets/sigmaaldrich/product/structures/381/265/db6ad39a-aa70-4869-b51d-64d96ea91c63/640/db6ad39a-aa70-4869-b51d-64d96ea91c63.png)
Sodium deoxycholate monohydrate
![Sodium deoxycholate monohydrate BioUltra, ≥99.0% (NT)](/deepweb/assets/sigmaaldrich/product/structures/381/265/db6ad39a-aa70-4869-b51d-64d96ea91c63/640/db6ad39a-aa70-4869-b51d-64d96ea91c63.png)
Synonym(s):
3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium salt, Desoxycholic acid sodium salt
Empirical Formula (Hill Notation):
C24H39NaO4 · H2O
Products
Shipping
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing