Skip to Content
Merck
Sodium 2-methyl-2-propanethiolate technical grade, 90%

Sodium 2-methyl-2-propanethiolate

Sodium 2-methyl-2-propanethiolate technical grade, 90%
Synonym(s):
2-Methyl-2-propanethiol sodium salt, Sodium tert-butyl sulfide, t-Butanethiol sodium salt
Linear Formula:
(CH3)3CSNa
CAS Number:
Molecular Weight:
112.17

Products

Shipping
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing