
Propofol impurity J

Synonym(s):
2,6-Diisopropyl-1,4-benzoquinone
Empirical Formula (Hill Notation):
C12H16O2
CAS Number:
Molecular Weight:
192.25
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing