![PCB No 209 solution 10±0.5 μg/mL in isooctane, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/368/551/1047ffbc-02b6-440c-868c-86a17d1a9054/640/1047ffbc-02b6-440c-868c-86a17d1a9054.png)
PCB No 209 solution
![PCB No 209 solution 10±0.5 μg/mL in isooctane, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/368/551/1047ffbc-02b6-440c-868c-86a17d1a9054/640/1047ffbc-02b6-440c-868c-86a17d1a9054.png)
Synonym(s):
Decachlorobiphenyl solution
Empirical Formula (Hill Notation):
C12Cl10
CAS Number:
Molecular Weight:
498.66
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing